1,1-Dimethoxyacetone ≥97%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
1,1-Dimethoxyacetone ≥97%
Catalog # B24988.22
Supplier: Thermo Fisher Scientific
CAS Number: 6342-56-9
Jump to:
Product Details & Documents
Purity: 97+% CAS No.: 6342-56-9 Molecular Formula: CH3COCH(OCH3)2 Formula Weight: 118.13 Flash Point: 34°(93°F) Refractive Index: 1.3980 Melting Point: -57° Boiling Point: 133-135° Beilstein Reference: 1560557 UN No.: 1993 MDL No.: MFCD00008758
Synonyms: Pyruvaldehyde 1,1-dimethyl acetal, 1,1-Dimethoxypropan-2-one, Pyruvic aldehyde dimethyl acetal
- CAS Number:6342-56-9
- Formula:C₅H₁₀O₃
- MDL Number:MFCD00008758
- Molecular Weight:118.13 g/mol
- Boiling Pt:133…135 °C
- Melting Pt:–57 °C
- Density:0.998
- Flash Pt:34 °C (93 °F)
- Storage Temperature:Ambient
- EINECS:228-735-4
- UN:1993
- ADR:3,III
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



