Order Entry
Italy
Orders LinkContactUsLinkComponent
 

2-Hydroxy-1-naphthaldehyde 98%

Glass bottle for solids, 100 g
Catalog # B23951.22
Irritant
2-Hydroxy-1-naphthaldehyde 98%
Catalog # B23951.22
CAS Number:  708-06-5
2-Hydroxy-1-naphthaldehyde 98%
2-Hydroxy-1-naphthaldehyde 98%

Glass bottle for solids, 100 g

CAS Number:  708-06-5
Price
115,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 98% CAS No.: 708-06-5 Molecular Formula: C11H8O2 Formula Weight: 172.18 Odour: Odourless Melting Point: 77-80° Boiling Point: 191-193°/27mm UN No.: 3077 MDL No.: MFCD00004005

Synonyms:  2-Hydroxy-α-naphthaldehyde
  • CAS Number:708-06-5
  • Formula:C₁₁H₈O₂
  • MDL Number:MFCD00004005
  • Molecular Weight:172.18 g/mol
  • Boiling Pt:191…193 °C (27 mmHg)
  • Melting Pt:77…80 °C
  • Storage Temperature:Ambient
  • EINECS:211-902-0
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications