Order Entry
Italy
Orders LinkContactUsLinkComponent
2-Hydroxy-1-naphthaldehyde 98%
Glass bottle for solids, 25 g
  B23951.14
2-Hydroxy-1-naphthaldehyde 98%
  B23951.14
 :  708-06-5
2-Hydroxy-1-naphthaldehyde 98%
2-Hydroxy-1-naphthaldehyde 98%
Glass bottle for solids, 25 g
 :  708-06-5
41,10 €
1 * 25 g
Purity: 98% CAS No.: 708-06-5 Molecular Formula: C11H8O2 Formula Weight: 172.18 Odour: Odourless Melting Point: 77-80° Boiling Point: 191-193°/27mm UN No.: 3077 MDL No.: MFCD00004005
 :  2-Hydroxy-α-naphthaldehyde
  •  :708-06-5
  •  :C₁₁H₈O₂
  •  :MFCD00004005
  •  :172.18 g/mol
  •  :191…193 °C (27 mmHg)
  •  :77…80 °C
  •  :Ambient
  •  :211-902-0
  •  :9,III
  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g