Order Entry
Italy
Orders LinkContactUsLinkComponent
 

6-Methoxy-2-naphthaldehyde ≥99%

Glass bottle for solids, 25 g
Catalog # B23943.14
6-Methoxy-2-naphthaldehyde ≥99%
Catalog # B23943.14
CAS Number:  3453-33-6
6-Methoxy-2-naphthaldehyde ≥99%
6-Methoxy-2-naphthaldehyde ≥99%

Glass bottle for solids, 25 g

CAS Number:  3453-33-6
Price
140,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 99% CAS No.: 3453-33-6 Molecular Formula: C12H10O2 Formula Weight: 186.21 Melting Point: 80-82° Beilstein Reference: 1945989

  • CAS Number:3453-33-6
  • Formula:C₁₂H₁₀O₂
  • MDL Number:MFCD00081152
  • Molecular Weight:186.21 g/mol
  • Melting Pt:80…82 °C
  • Storage Temperature:Ambient
  • EINECS:222-377-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications