Order Entry
Italy
Orders LinkContactUsLinkComponent
Daidzein ≥97%

Daidzein ≥97%

Glass bottle for solids, 1 g
Catalog # B22877.03
Irritant
Daidzein ≥97%
Catalog # B22877.03
CAS Number:  486-66-8
Daidzein ≥97%
Daidzein ≥97%

Glass bottle for solids, 1 g

CAS Number:  486-66-8
Price
57,00 €
Log in for availability
1 * 1 g
Product Details & Documents

Purity: 97% CAS No.: 486-66-8 Molecular Formula: C15H10O4 Formula Weight: 254.24 Melting Point: 328-332° dec. Beilstein Reference: 231523 Merck Reference: 14,2801 MDL No.: MFCD00016954

Synonyms:  Daidzeol, 7-Hydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 7-Hydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',7-Dihydroxyisoflavone, 7-Hydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one
  • CAS Number:486-66-8
  • Formula:C₁₅H₁₀O₄
  • MDL Number:MFCD00016954
  • Molecular Weight:254.24 g/mol
  • Merck Index:14,2801
  • Boiling Pt:513 °C (1013 hPa)
  • Melting Pt:328…332 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:207-635-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.