Inserisci ordine
Italy
Orders LinkContactUsLinkComponent
 

1-(4-Morpholino)cyclohexene ≥97%

Glass bottle for solids, 100 g
Catalog # B22004.22
Irritant
1-(4-Morpholino)cyclohexene ≥97%
Catalog # B22004.22
CAS Number:  670-80-4
1-(4-Morpholino)cyclohexene ≥97%
1-(4-Morpholino)cyclohexene ≥97%

Glass bottle for solids, 100 g

CAS Number:  670-80-4
Price
95,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 97% CAS No.: 670-80-4 Molecular Formula: C10H17NO Formula Weight: 167.25 Storage Temperature: 2°C to 8°C Flash Point: 68°(154°F) Refractive Index: 1.5146 Boiling Point: 118-120°/10mm Beilstein Reference: 118696 MDL No.: MFCD00006163

Synonyms:  N-(1-Cyclohexen-1-yl)morpholine, 1-Morpholinocyclohexene, 4-(1-Cyclohexen-1-yl)morpholine
  • CAS Number:670-80-4
  • Formula:C₁₀H₁₇NO
  • MDL Number:MFCD00006163
  • Molecular Weight:167.25 g/mol
  • Boiling Pt:118…120 °C (10 mmHg)
  • Density:1,020
  • Flash Pt:68 °C (154 °F)
  • EINECS:211-579-6
Specifications

Specifications

  • Tipo di conf.:
    Glass bottle for solids
  • Conf:
    100 g

Specifications