Order Entry
Italy
Orders LinkContactUsLinkComponent
1-(4-Morpholino)cyclohexene ≥97%
Glass bottle for solids, 25 g
  B22004.14
1-(4-Morpholino)cyclohexene ≥97%
  B22004.14
 :  670-80-4
1-(4-Morpholino)cyclohexene ≥97%
1-(4-Morpholino)cyclohexene ≥97%
Glass bottle for solids, 25 g
 :  670-80-4
33,90 €
1 * 25 g
Purity: 97% CAS No.: 670-80-4 Molecular Formula: C10H17NO Formula Weight: 167.25 Storage Temperature: 2°C to 8°C Flash Point: 68°(154°F) Refractive Index: 1.5146 Boiling Point: 118-120°/10mm Beilstein Reference: 118696 MDL No.: MFCD00006163
 :  N-(1-Cyclohexen-1-yl)morpholine, 1-Morpholinocyclohexene, 4-(1-Cyclohexen-1-yl)morpholine
  •  :670-80-4
  •  :C₁₀H₁₇NO
  •  :MFCD00006163
  •  :167.25 g/mol
  •  :118…120 °C (10 mmHg)
  •  :1.020
  •  :68 °C (154 °F)
  •  :211-579-6
  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g