Order Entry
Italy
Orders LinkContactUsLinkComponent
3,4-Dihydroxybenzhydrazide ≥97%

3,4-Dihydroxybenzhydrazide ≥97%

Glass bottle for solids, 25 g
Catalog # B21746.14
Irritant
3,4-Dihydroxybenzhydrazide ≥97%
Catalog # B21746.14
CAS Number:  39635-11-5
3,4-Dihydroxybenzhydrazide ≥97%
3,4-Dihydroxybenzhydrazide ≥97%

Glass bottle for solids, 25 g

CAS Number:  39635-11-5
Price
236,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 97% CAS No.: 39635-11-5 Molecular Formula: C7H8N2O3 Formula Weight: 168.15 Melting Point: ca 265° dec. Beilstein Reference: 2642968

Synonyms:  3,4-Dihydroxybenzoic hydrazide, 3,4-Dihydroxybenzoic acid hydrazide, 3,4-Dihydroxybenzene-1-carbohydrazide, 3,4-Dihydroxybenzohydrazide
  • CAS Number:39635-11-5
  • Formula:C₇H₈N₂O₃
  • MDL Number:MFCD00017064
  • Molecular Weight:168.15 g/mol
  • Melting Pt:∼ 265 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:254-550-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.