Order Entry
Italy
Orders LinkContactUsLinkComponent
3,4-Dihydroxybenzhydrazide ≥97%

3,4-Dihydroxybenzhydrazide ≥97%

Glass bottle for solids, 5 g
Catalog # B21746.06
Irritant
3,4-Dihydroxybenzhydrazide ≥97%
Catalog # B21746.06
CAS Number:  39635-11-5
3,4-Dihydroxybenzhydrazide ≥97%
3,4-Dihydroxybenzhydrazide ≥97%

Glass bottle for solids, 5 g

CAS Number:  39635-11-5
Price
61,50 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 97% CAS No.: 39635-11-5 Molecular Formula: C7H8N2O3 Formula Weight: 168.15 Melting Point: ca 265° dec. Beilstein Reference: 2642968

Synonyms:  3,4-Dihydroxybenzoic hydrazide, 3,4-Dihydroxybenzoic acid hydrazide, 3,4-Dihydroxybenzene-1-carbohydrazide, 3,4-Dihydroxybenzohydrazide
  • CAS Number:39635-11-5
  • Formula:C₇H₈N₂O₃
  • MDL Number:MFCD00017064
  • Molecular Weight:168.15 g/mol
  • Melting Pt:∼ 265 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:254-550-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications