Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Thioacetamide ≥98%

Glass bottle for solids, 100 g
Catalog # A12926.22
Health
Irritant
Thioacetamide ≥98%
Catalog # A12926.22
CAS Number:  62-55-5
Thioacetamide ≥98%
Thioacetamide ≥98%

Glass bottle for solids, 100 g

CAS Number:  62-55-5
Price
79,50 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 98% CAS No.: 62-55-5 Molecular Formula: C2H5NS Formula Weight: 75.13 Melting Point: 110-114° Solubility: Soluble in water, ethanol Beilstein Reference: 506006 Merck Reference: 14,9319 MDL No.: MFCD00008070

Synonyms:  Ethanethioamide
  • CAS Number:62-55-5
  • Formula:CH₃CSNH₂
  • MDL Number:MFCD00008070
  • Molecular Weight:75.13 g/mol
  • Merck Index:14,9319
  • Melting Pt:110…114 °C
  • Density:1.37
  • Storage Temperature:Ambient
  • EINECS:200-541-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.