1,6-Hexanediol ≥97%
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 97% CAS No.: 629-11-8 Molecular Formula: HO(CH2)6OH Formula Weight: 118.18 Flash Point: 147°(296°F) Physical Form: Cryst solid Density: 1.120 Sensitivity: Hygroscopic Appearance: White Melting Point: 40-43° Boiling Point: 248-250° Beilstein Reference: 1633461 Merck Reference: 14,4690 MDL No.: MFCD00002985
Synonyms: Hexamethylene glycol, 1,6-Dihydroxy hexane
- CAS Number:629-11-8
- Formula:C₆H₁₄O₂
- MDL Number:MFCD00002985
- Molecular Weight:118.18 g/mol
- Merck Index:14,4690
- Boiling Pt:248…250 °C
- Melting Pt:40…43 °C
- Density:1.120
- Flash Pt:147 °C (296 °F)
- Storage Temperature:Ambient
- EINECS:211-074-0
Documents
Certificates
Search by Lot NumberALFAA12439.30_TDS_EN
Product specification
Organic Building Blocks and Intermediates
Application Notes
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



