Order Entry
Italy
Orders LinkContactUsLinkComponent
 

2,6-Dibromotoluene ≥98%

Glass bottle for solids, 1 g
Codice catalogo A11832.03
Irritant
Nome del fornitore:   Thermo Fisher Scientific
2,6-Dibromotoluene ≥98%
Codice catalogo A11832.03
Nome del fornitore:  Thermo Fisher Scientific
Numero CAS:  69321-60-4
2,6-Dibromotoluene ≥98%
2,6-Dibromotoluene ≥98%

Glass bottle for solids, 1 g

Numero CAS:  69321-60-4
Price
40,10 €
Log in for availability
1 * 1 g
Product Details & Documents

Purity: 98+% CAS No.: 69321-60-4 Molecular Formula: C7H6Br2 Formula Weight: 249.94 Refractive Index: 1.6060 Melting Point: 2-6° Boiling Point: 112-113°/7mm Beilstein Reference: 3235502 MDL No.: MFCD00013524

  • CAS Number:69321-60-4
  • Formula:C₇H₆Br₂
  • MDL Number:MFCD00013524
  • Molecular Weight:249.93 g/mol
  • Boiling Pt:112…113 °C (7 mmHg)
  • Melting Pt:2…6 °C
  • Density:1.812
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications