Order Entry
Italy
Orders LinkContactUsLinkComponent
Lithium fluoride 97%

Lithium fluoride 97%

Glass bottle for solids, 100 g
Catalog # A11632.22
Toxic
Irritant
Lithium fluoride 97%
Catalog # A11632.22
CAS Number:  7789-24-4
Lithium fluoride 97%
Lithium fluoride 97%

Glass bottle for solids, 100 g

CAS Number:  7789-24-4
Price
49,40 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 98+% CAS No.: 7789-24-4 Molecular Formula: LiF Formula Weight: 25.94 Refractive Index: 1.3915 Odour: Odourless Boiling Point: 1680° Merck Reference: 14,5531 UN No.: 3288 MDL No.: MFCD00011090

  • CAS Number:7789-24-4
  • Formula:FLi
  • MDL Number:MFCD00011090
  • Molecular Weight:25.94 g/mol
  • Merck Index:14,5531
  • Boiling Pt:1680 °C
  • Melting Pt:845 °C
  • Density:2.635
  • Storage Temperature:Ambient
  • EINECS:232-152-0
  • UN:3288
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.