Soda lime, with indicator
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Soda lime, with indicator
Catalog # 44786.36
Supplier: Thermo Fisher Scientific
CAS Number: 8006-28-8
Jump to:
Product Details & Documents
CAS No.: 8006-28-8 Physical Form: 8-12 Mesh Sensitivity: Air Sensitive and Hygroscopic Odour: Odourless Merck Reference: 14,8569 UN No.: 1907 MDL No.: MFCD00134094
- CAS Number:8006-28-8
- MDL Number:MFCD00134094
- Merck Index:14,8569
- Storage Temperature:Ambient
- UN:1907
- ADR:8,III
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



