Order Entry
Italy
Orders LinkContactUsLinkComponent
2-Aminoadenosine ≥97%

2-Aminoadenosine ≥97%

25 g
Catalog # 44748.14
2-Aminoadenosine ≥97%
Catalog # 44748.14
CAS Number:  2096-10-8
2-Aminoadenosine ≥97%
2-Aminoadenosine ≥97%

25 g

CAS Number:  2096-10-8
Price
552,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 97% CAS No.: 2096-10-8 Molecular Formula: C10H14N6O4 Formula Weight: 282.26 Physical Form: Powder MDL No.: MFCD00053556

Synonyms:  2,6-Diamino-9-(beta-D-ribofuranosyl)purine, (2R,3R,4S,5R)-2-(2,6-Diamino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, 2,6-Diaminopurine Riboside
  • CAS Number:2096-10-8
  • Formula:C₁₀H₁₄N₆O₄
  • MDL Number:MFCD00053556
  • Molecular Weight:282.26 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.