Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Cylinders, without lid

100 mm, 50 mm
Catalog # HECH44440014
Cylinders, without lid
Catalog # HECH44440014
Cylinders, without lid
Cylinders, without lid

100 mm, 50 mm

Price
26,20 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

Jars, made of glass or stainless steel, with base, without lids.

Suitable for storing dressing material, cotton, clinical thermometers, forceps and other instruments.

 : Lids with knob available must be ordered separately.

Specifications
  • Ext.Ø:
    50 mm
  • Height:
    100 mm
  • Seq:
    12