Order Entry
Italy
Orders LinkContactUsLinkComponent
Additional Top and Bottom Shelves
Top shelf, perforated, for cleanroom heavy duty tables, W×D×H: 900×300×700, fabricated from stainless steel 304 with electropolished finish
  132-0379
 :   Palbam
Additional Top and Bottom Shelves
  132-0379
 :  Palbam
Additional Top and Bottom Shelves
Additional Top and Bottom Shelves
Top shelf, perforated, for cleanroom heavy duty tables, W×D×H: 900×300×700, fabricated from stainless steel 304 with electropolished finish
1 * 1 pezzi
  • Description:
    Top shelf, perforated, for cleanroom heavy duty tables, W×D×H: 900×300×700, fabricated from stainless steel 304 with electropolished finish
  • Seq:
    15