Order Entry
Italy
Orders LinkContactUsLinkComponent
Screw caps
Screw caps
GL 40
Catalog # VITL83315
Supplier:   VITLAB
Screw caps
Catalog # VITL83315
Supplier:  VITLAB
Screw caps
Screw caps
GL 40
Price
30,40 €
Log in for availability
1 * 12 pezzi
Product Details & Documents
Made of PP, translucent.
Caps are autoclavable at 121 °C (2 bar) according to DIN EN 285.
Specifications
  • Thread:
    GL 40