Screw caps
GL 18
Catalog # VITL83310
Supplier: VITLAB
Jump to:
Product Details & Documents
Made of PP, translucent.
Caps are autoclavable at 121 °C (2 bar) according to DIN EN 285.
Specifications
- Thread:GL 18