Inserisci ordine
Italy
Orders LinkContactUsLinkComponent
Tappi, in PPCO, profilo basso, 11 mm

Tappi, in PPCO, profilo basso, 11 mm

porpora, 11 mm, -
Catalog # 215-1654
   Produzione a basso impatto
Tappi, in PPCO, profilo basso, 11 mm
Catalog # 215-1654
   Produzione a basso impatto
Tappi, in PPCO, profilo basso, 11 mm
Tappi, in PPCO, profilo basso, 11 mm

porpora, 11 mm, -

Price
173,00 €
Log in for availability
1 * 1.000 pezzi
Product Details & Documents

Identify samples quickly with low profile closures for micro packaging vials. Polypropylene polymer closures are ready to use.

  • RNase/DNase free (only sterile closures), non-cytotoxic and non-pyrogenic

Certifications: Manufactured in compliance with ISO 13485:2003 quality requirements. Micro packaging vials and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.

Packaging: Closures are bulk packed without vials, 500 per inner bag, 1000 per case.

Specifications
  • Dimensione tappo:
    11 mm
  • Sterili:
    -
  • Seq:
    15
  • Colore tappo:
    porpora
  • Cat. No.:
    215-1654
  • Environmentally Preferable:
    Produzione a basso impatto
  • Certificazione ambientale:
    NAR Certificate of Retirement