Order Entry
Italy
Orders LinkContactUsLinkComponent
Closures, PPCO, low profile, 11 mm

Closures, PPCO, low profile, 11 mm

Natural, 11 mm, -
Codice catalogo 215-1651
   Low Manufacturing Impact
Nome del fornitore:   Thermo Fisher Scientific
Closures, PPCO, low profile, 11 mm
Codice catalogo 215-1651
Nome del fornitore:  Thermo Fisher Scientific
   Low Manufacturing Impact
Closures, PPCO, low profile, 11 mm
Closures, PPCO, low profile, 11 mm

Natural, 11 mm, -

Price
173,00 €
Log in for availability
1 * 1.000 pezzi
Product Details & Documents

Identify samples quickly with low profile closures for micro packaging vials. Polypropylene polymer closures are ready to use.

  • RNase/DNase free (only sterile closures), non-cytotoxic and non-pyrogenic

Certifications: Manufactured in compliance with ISO 13485:2003 quality requirements. Micro packaging vials and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.

Packaging: Closures are bulk packed without vials, 500 per inner bag, 1000 per case.

Specifications
  • Cap Size:
    11 mm
  • Sterile:
    -
  • Seq:
    09
  • Cap colour:
    Natural
  • Cat. No.:
    215-1651
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact