Order Entry
Italy
Orders LinkContactUsLinkComponent
Wipes for general applications, Tork

Wipes for general applications, Tork

Combi roll, for use with W1/W2/W3 dispensers, White, 152 m × 320 mm, Centrefeed roll of 400 sheets
Catalog # 115-0338
Supplier:   Essity
Wipes for general applications, Tork
Catalog # 115-0338
Supplier:  Essity
Wipes for general applications, Tork
Wipes for general applications, Tork

Combi roll, for use with W1/W2/W3 dispensers, White, 152 m × 320 mm, Centrefeed roll of 400 sheets

Price
61,40 €
Log in for availability
1 * 1 Roll
Product Details & Documents

Strong and durable cloth wipes for use in a variety of environments, softness and flexibility mean wipes are excellent for cleaning, wiping and polishing even in confined spaces.

  • Cellulose/PP/polyester, reusable
  • Solvent resistant, good dry and wet strength and performance
  • High absorbency
  • Low-linting

Colour: White or blue.

Colour: White or blue

Specifications
  • Packed:
    Centrefeed roll of 400 sheets
  • Colour:
    White
  • Description:
    Combi roll, for use with W1/W2/W3 dispensers
  • L×W:
    152 m × 320 mm
  • Seq:
    01
Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..