Order Entry
Italy
Orders LinkContactUsLinkComponent
 

3-Mercaptopropionic acid ≥99%

Glass bottle for solids, 500 g
Codice catalogo A13261.36
Toxic
Corrosive
Nome del fornitore:   Thermo Fisher Scientific
3-Mercaptopropionic acid ≥99%
Codice catalogo A13261.36
Nome del fornitore:  Thermo Fisher Scientific
Numero CAS:  107-96-0
3-Mercaptopropionic acid ≥99%
3-Mercaptopropionic acid ≥99%

Glass bottle for solids, 500 g

Numero CAS:  107-96-0
Price
93,60 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99% CAS No.: 107-96-0 Molecular Formula: C3H6O2S Formula Weight: 106.14 Flash Point: 93°(199°F) Refractive Index: 1.4920 Melting Point: 17-19° Boiling Point: 110-111°/15mm Beilstein Reference: 773807 UN No.: 2922 MDL No.: MFCD00004897

  • CAS Number:107-96-0
  • Formula:C₃H₆O₂S
  • MDL Number:MFCD00004897
  • Molecular Weight:106.15 g/mol
  • Boiling Pt:110…111 °C (15 mmHg)
  • Melting Pt:17…19 °C
  • Density:1.220
  • Flash Pt:93 °C (199 °F)
  • Storage Temperature:Ambient
  • EINECS:203-537-0
  • UN:2922
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.