Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Daidzein

Glass bottle for solids, 500 mg
Catalog # CAYM10005166-500
Supplier:   Cayman Chemical
Daidzein
Catalog # CAYM10005166-500
Supplier:  Cayman Chemical
CAS Number:  486-66-8
Daidzein

Glass bottle for solids, 500 mg

CAS Number:  486-66-8
Price
97,20 €
Log in for availability
1 * 500 mg
Product Details & Documents
Synonyms:  Daidzeol, 7-Hydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 7-Hydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',7-Dihydroxyisoflavone, 7-Hydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one
  • CAS Number:486-66-8
  • Formula:C₁₅H₁₀O₄
  • MDL Number:MFCD00016954
  • Molecular Weight:254.24 g/mol
  • Boiling Pt:513 °C (1013 hPa)
  • Melting Pt:212…214 °C
  • EINECS:207-635-4

 

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg

Specifications