Powder flasks with conical ground joint
250 ml, 29/32 NS
Catalog # 201-0626
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN®, borosilicate glass, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Especially for rotary evaporators, for drying of powder-like substances. With center neck, conical joint and inscription label.
Specifications
- Socket size:29/32 NS
- Capacity:250 ml