Order Entry
Italy
Orders LinkContactUsLinkComponent
VISOCOLOR® Colorimetric Test, Ammonia

VISOCOLOR® Colorimetric Test, Ammonia

VISOCOLOR® ECO Ammonium 15, refill pack, test no. 5-10; Measuring range on PF-12Plus. Range on other photometers can be different, Reagents for 50 tests in a box, 0 • 0,5 • 1 • 2 • 3 • 5 • 7 • 10 • 15 mg/L (NH₄⁺); 0,5 - 8,0 mg/L (NH₄⁺), Indophenol, PF-12Plus; Advance; Visual, 170×115×70 mm (Including packaging), Sea water compatible, 50, 1 KIT
Catalog # MANA931210
Supplier:   MACHEREY-NAGEL
VISOCOLOR® Colorimetric Test, Ammonia
Catalog # MANA931210
Supplier:  MACHEREY-NAGEL
CAS Number:  
VISOCOLOR® Colorimetric Test, Ammonia
VISOCOLOR® Colorimetric Test, Ammonia

VISOCOLOR® ECO Ammonium 15, refill pack, test no. 5-10; Measuring range on PF-12Plus. Range on other photometers can be different, Reagents for 50 tests in a box, 0 • 0,5 • 1 • 2 • 3 • 5 • 7 • 10 • 15 mg/L (NH₄⁺); 0,5 - 8,0 mg/L (NH₄⁺), Indophenol, PF-12Plus; Advance; Visual, 170×115×70 mm (Including packaging), Sea water compatible, 50, 1 KIT

CAS Number:  
Price
44,50 €
Log in for availability
1 * 1 KIT
Specifications

Specifications

  • Chemical method:
    Indophenol
  • Pk:
    1 KIT
  • Description:
    VISOCOLOR® ECO Ammonium 15, refill pack, test no. 5-10; Measuring range on PF-12Plus. Range on other photometers can be different
  • Dimensions:
    170×115×70 mm (Including packaging)
  • Type:
    Sea water compatible
  • For unit:
    PF-12Plus; Advance; Visual
  • Tests per kit:
    50
  • Range:
    0 • 0,5 • 1 • 2 • 3 • 5 • 7 • 10 • 15 mg/L (NH₄⁺); 0,5 - 8,0 mg/L (NH₄⁺)
  • Contents:
    Reagents for 50 tests in a box
  • Cat. No.:
    MANA931210
  • Platform:
    Colorimetric test kits
  • Model:
    MANA931210
  • Shelf life (months):
    18
  • Storage temperature:
    15...25 °C
  • Weight:
    130 g (Including packaging)

Specifications