Sleeves for conical joints
24/29
Catalog # KART1040
Supplier: KARTELL
Jump to:
Product Details & Documents
PTFE.
- Very good chemical resistance
- Gas, liquid and vacuum-tight (to 0,1 mbar)
- Temperature resistant from −200 to +280 °C
For fitting onto standard ground cones. Easily detachable and suitable for repeated use. Joint grease is not needed.
Specifications
- For NS:24/29
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



