Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Buffers
Biological Buffers
RNA isolation buffers
Print
RNA isolation buffers
TRK lysis buffer, 100 mL
Catalog # PR021
SDS
Supplier:
OMEGA BIO-TEK
RNA isolation buffers
Catalog # PR021
Supplier:
OMEGA BIO-TEK
CAS Number:
SDS
Return to Top
RNA isolation buffers
TRK lysis buffer, 100 mL
Specifications
Jump to:
Specifications
CAS Number:
Price
186,00 €
Log in
for availability
1 * 100 mL
-
+
Other Product Family Options Available
Return Policy
Product Details & Documents
sds_VWRCPR031
Specifications
Specifications
Pk:
100 mL
Description:
TRK lysis buffer
Specifications
VWR®, PCR Plates, 96 Well
#: 732-3388
Supplier: VWR Collection
Microscope slides, Menzel Gläser
#: 630-1985
Supplier: Epredia