Order Entry
Italy
Orders LinkContactUsLinkComponent
Standard ground joints
Standard ground joints
- NS, 14/23 NS, 17 mm
Catalog # QUTW1030
Standard ground joints
Catalog # QUTW1030
Standard ground joints
Standard ground joints
- NS, 14/23 NS, 17 mm
Price
102,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
Connections, quartz glass, with standard ground joint according to DIN 12 249.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving high resistance to temperature changes.
Specifications
  • Cone size:
    - NS
  • Socket size:
    14/23 NS
  • Tube ext.Ø:
    17 mm
  • Seq:
    01