Standard ground joints
29/32 NS, - NS, 26 mm
Catalog # QUTW1047
Supplier: QUARTZTECHNISCHE WERKSTATTEN
Jump to:
Product Details & Documents
Connections, quartz glass, with standard ground joint according to DIN 12 249.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving high resistance to temperature changes.
Specifications
- Cone size:29/32 NS
- Socket size:- NS
- Tube ext.Ø:26 mm
- Seq:09