Order Entry
Italy
ContactUsLinkComponent
Stopper, rubber, grey, diameter(bottomx top)41x49mm, height 40mm 1 drilling diameter. 9mm. 1 * 1 items
Catalog # DENE101194101/9
Supplier:  DEUTSCH NEUMANN
undefined
Stopper, rubber, grey, diameter(bottomx top)41x49mm, height 40mm 1 drilling diameter. 9mm. 1 * 1 items
Catalog # DENE101194101/9
Supplier:  DEUTSCH NEUMANN