Order Entry
Italy
Orders LinkContactUsLinkComponent
Beakers, low form, with spout
100 ml, 70 mm, 50 mm
  WITG8.600.100
Beakers, low form, with spout
  WITG8.600.100
Beakers, low form, with spout
Beakers, low form, with spout
100 ml, 70 mm, 50 mm
1 * 1 pezzi
Quartz glass.
  • High thermal shock resistance with low thermal expansion
  • High transmission from the UV to the IR spectrum range
  • High chemical resistance and purity
  • High electrical insulation properties
  • High radiation resistance
  • High temperature resistance and softening temperature
The combination of the special thermal, electrical, chemical, mechanical and optical properties in a single material makes quartz glass indispensible for high-technology applications.
  • Ext.Ø:
    50 mm
  • Capacity:
    100 ml
  • Height:
    70 mm