Order Entry
Italy
Orders LinkContactUsLinkComponent
Pipette tips, universal, Axygen®
Pipette tips, universal, Axygen®
Standard, 0,5 - 10 µl, -, Transparent, 10 racks, 96 each
Catalog # 613-0641
Supplier:   Corning
Pipette tips, universal, Axygen®
Catalog # 613-0641
Supplier:  Corning
Pipette tips, universal, Axygen®
Pipette tips, universal, Axygen®
Standard, 0,5 - 10 µl, -, Transparent, 10 racks, 96 each
Price
56,70 €
Log in for availability
1 * 960 pezzi
Product Details & Documents
These universal pipette tips are compatible with most micropipettes.
  • PP, autoclavable
  • Smooth finish for minimum retention of collected sample
  • Certified free of RNase, DNase and endotoxins
  • Maxymum Recovery® tips feature ultra smooth surfaces: For reduced sample loss and maximal accuracy and precision
Ordering information: Available loose or in racks, sterile or non sterile versions.
Specifications
  • Packed:
    10 racks, 96 each
  • Sterile:
    -
  • Volume:
    0,5 - 10 µl
  • Tip style:
    Standard
  • Tip colour:
    Transparent