Order Entry
Italy
ContactUsLinkComponent
FRITSCH-test powder F-500, 0.5 - 50µm, for wet and dry dispersion, (50 g) for Laser Particle Sizer ANALYSETTE 22 NanoTec For calibration of the Laser Particle Sizer ANALYSETTE 22 NanoTec for wet and dry dispersion within the framework of a test equip 1 * 1 items
Catalog # FRIT85.2100.00
Supplier:  FRITSCH
FRITSCH-test powder F-500, 0.5 - 50µm, for wet and dry dispersion, (50 g) for Laser Particle Sizer ANALYSETTE 22 NanoTec For calibration of the Laser Particle Sizer ANALYSETTE 22 NanoTec for wet and dry dispersion within the framework of a test equip 1 * 1 items
Catalog # FRIT85.2100.00
Supplier:  FRITSCH
Frequently Bought Together