Thermometer, maxi T, digital, electronic, measuring range: -10:200/0.1C, piercing probe of stainless steel 125x3.5mm, switchable F , maximum/minimum function, hold-function, water-resistant. 1 * 1 items
Catalog # AMARE905180
Nome del fornitore: Amarell
Frequently Bought Together