Order Entry
Italy
Orders LinkContactUsLinkComponent
Affinity chromatography columns, GSTrap™ HP

Affinity chromatography columns, GSTrap™ HP

GSTrap™ HP, PP, 34 µm, 16×25 mm, 5×5 ml, <20 ml/min, <600 cm/h
Catalog # 17-5282-02
Supplier:   Cytiva
Affinity chromatography columns, GSTrap™ HP
Catalog # 17-5282-02
Supplier:  Cytiva
Affinity chromatography columns, GSTrap™ HP
Affinity chromatography columns, GSTrap™ HP

GSTrap™ HP, PP, 34 µm, 16×25 mm, 5×5 ml, <20 ml/min, <600 cm/h

Price
1 670,00 €
Log in for availability
1 * 5 pezzi
Product Details & Documents

GSTrap™ HP columns are prepacked with Glutathione Sepharose High Performance resin and provide high-resolution, one-step purifications of glutathione S-Transferase (GST) tagged proteins.

  • High resolution purification of GST-tagged proteins
  • Mild elution conditions preserving protein antigenicity and function
  • Fast, one-step purification
  • Easy to use with syringe, pump or a chromatography system

GSTrap™ HP 1-ml and 5-ml columns are prepacked with Glutathione Sepharose High Performance resin, which provides high-resolution purifications, thus a more concentrated sample. GST-tagged proteins are purified from pretreated bacterial lysates by affinity chromatography using immobilized glutathione, such as Glutathione Sepharose High Performance. GST-tagged proteins are bound to the affinity resin, and impurities are removed by washing with binding buffer

GST-tagged proteins are eluted under mild, nondenaturing conditions using reduced glutathione. The purification process preserves protein antigenicity and function. If desired, cleavage of the protein from GST can be achieved using a site-specific protease whose recognition sequence is located immediately upstream from the multiple cloning site on the pGEX plasmids. GST-tagged proteins can be detected using colorimetric or immunological methods.

Specifications
  • Flow rate:
    <20 ml/min
  • Particle Size:
    34 µm
  • Capacity:
    5×5 ml
  • Description:
    GSTrap™ HP
  • Seq:
    04
  • Column hardware:
    PP
  • Bed dimensions:
    16×25 mm
  • Flow velocity:
    <600 cm/h
  • Cat. No.:
    17-5282-02
  • Max. pressure:
    5 bar
  • Storage:
    4 to 30 °C, 20% Ethanol
  • pH range:
    3 - 12