VISOCOLOR® Colorimetric Test, Ammonia
VISOCOLOR® ECO Ammonium 15, test no. 5-10; Measuring range on PF-12Plus. Range on other photometers can be different, Reagents for 50 tests, colour chart and accessories in a box, 0 • 0,5 • 1 • 2 • 3 • 5 • 7 • 10 • 15 mg/L (NH₄⁺); 0,5 - 8,0 mg/L (NH₄⁺), Indophenol, PF-12Plus; Advance; Visual, 170×125×45 mm (Including packaging), Sea water compatible, 50, 1 KIT
Supplier: MACHEREY-NAGEL
Jump to:
Product Details & Documents
Reagents for the determination of ammonia in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Hazardous material: Yes
Documents
Certificates
Search by Lot NumberIFU VISOCOLOR® Alpha Ammonium NH₄⁺.pdf IFU VISOCOLOR® ECO Ammonium 15.pdf IFU VISOCOLOR® ECO Ammonium 3.pdf IFU VISOCOLOR® HE Ammonium.pdf IFU_Ammonium 15 ECO Visocolor
More
Specifications
Specifications
- Chemical method:Indophenol
- Pk:1 KIT
- Description:VISOCOLOR® ECO Ammonium 15, test no. 5-10; Measuring range on PF-12Plus. Range on other photometers can be different
- Dimensions:170×125×45 mm (Including packaging)
- Type:Sea water compatible
- For unit:PF-12Plus; Advance; Visual
- Tests per kit:50
- Range:0 • 0,5 • 1 • 2 • 3 • 5 • 7 • 10 • 15 mg/L (NH₄⁺); 0,5 - 8,0 mg/L (NH₄⁺)
- Contents:Reagents for 50 tests, colour chart and accessories in a box
- Cat. No.:MANA931010
- Platform:Colorimetric test kits
- Model:MANA931010
- Shelf life (months):18
- Storage temperature:15...25 °C
- Weight:226,8 g (Including packaging)
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



