Order Entry
Italy
Orders LinkContactUsLinkComponent
BD Microlance™ 3 Needles
BD Microlance™ 3 Needles
Catalog # 613-3896
BD Microlance™ 3 Needles
Catalog # 613-3896
Supplier:  
BD Microlance™ 3 Needles
BD Microlance™ 3 Needles
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Kanüle 27G 1/2 - 0,4 x 12,7 mm #N/A #N/ Catalog #613-3832
Product 613-3832 is similar to 613-3896 but manufacturer and specifications may vary.
Product Details & Documents
The BD Microlance™ 3 needles are designed for easy penetration, smooth gliding force during insertion/withdraw, and reduced cannula friction while sliding through tissue. These needles are available in a wide range of gauges, lengths, and bevel designs.
  • Recommended use: intradermal, subcutaneous, intramuscular, intravenous injections, and aspiration
  • BD Microlance™ needles can be connected to a luer slip adapter or luer-lock connector
  • Natural rubber latex is not part of the material formulation
  • Sterile, single-use design
Conventional needles with a regular size wall are less likely to flex than thin wall needles, due to the added cannula thickness. A thin wall needles inner diameter potentially allows use of a needle gauge that is one size smaller than that of a regular needle, while benefiting from the same flow rate.
 : Individually packed in blister film and medical grade paper.