Screw caps for laboratory bottles
Screw cap, with lip seal, GL 45, giallo
Codice catalogo BIBB4506/45F
Nome del fornitore: SciLabware
Jump to:
Product Details & Documents
Screw cap, made of PP, for media-lab bottles
- Plug seal cap ideal for media preparation work
- Autoclavable up to 140°C
- Available in choice of 4 colours for easy identification of samples
Documents
Specifications
- Filettatura:GL 45
- Colore:giallo
- Descrizione:Screw cap, with lip seal
- Seq:05