Joint clips for conical joints, POM, Quickfit®
Ochre, 40/38
Codice catalogo 201-3032
Nome del fornitore: SciLabware
Joint clips for conical joints, POM, Quickfit®
Codice catalogo 201-3032
Nome del fornitore: SciLabware
Jump to:
Product Details & Documents
POM, coloured. Polyacetal clips for retaining glassware having Quickfit® conical joints.
- Easy to fit and remove
- Can be used at temperatures up to +80 °C
- Clips are colour coded for easy identification
Specifications
- For NS:40/38
- Colour:Ochre
- Seq:08