Accessories for screw thread joints
6,0 -7,0 mm, 13
Catalog # 201-0895
Supplier: SciLabware
Jump to:
Product Details & Documents
For use with Quickfit® glass screw thread.
Specifications
- Hole Ø:6,0 -7,0 mm
- Thread:13