Polysorbate 40
Glass bottle, 100 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 9005-66-7 Molecular Formula: C62H122O26 Refractive Index: 1.4700 MDL No.: MFCD00165345
Synonyms: Sorbitan monopalmitate, ethoxylated, Polyoxyethylene sorbitan monopalmitate
- CAS Number:9005-66-7
- Formula:C₆₂H₁₂₂O₂₆
- MDL Number:MFCD00165345
- Density:1.090
- Storage Temperature:Ambient
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:100 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



