Lithium-2-ethylhexanoate
Glass bottle for solids, 10 g
Supplier: Thermo Fisher Scientific
Lithium-2-ethylhexanoate
Catalog # 39330.09
Supplier: Thermo Fisher Scientific
CAS Number: 15590-62-2
Jump to:
Product Details & Documents
CAS No.: 15590-62-2 Molecular Formula: C8H15LiO2 Formula Weight: 150.15 Physical Form: Lump MDL No.: MFCD00058702
Synonyms: Lithium 2-ethylhexanoate, 2-Ethylhexanoic acid lithium salt, 2-Ethylcaproic acid lithium salt
- CAS Number:15590-62-2
- Formula:C₈H₁₅LiO₂
- MDL Number:MFCD00058702
- Molecular Weight:150.15 g/mol
- Storage Temperature:Ambient
- EINECS:239-657-5
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



