Order Entry
Italy
Orders LinkContactUsLinkComponent
 

1,6-Hexanedithiol ≥97%

Glass bottle for solids, 5 g
Catalog # L06686.06
1,6-Hexanedithiol ≥97%
Catalog # L06686.06
CAS Number:  1191-43-1
1,6-Hexanedithiol ≥97%
1,6-Hexanedithiol ≥97%

Glass bottle for solids, 5 g

CAS Number:  1191-43-1
Price
26,80 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 97% CAS No.: 1191-43-1 Molecular Formula: HS(CH2)6SH Formula Weight: 150.30 Flash Point: 90°(194°F) Refractive Index: 1.5130 Melting Point: -21° Boiling Point: 242-243° Beilstein Reference: 1732507 UN No.: 2810 MDL No.: MFCD00004910

Synonyms:  1,6-Dimercaptohexane
  • CAS Number:1191-43-1
  • Formula:C₆H₁₄S₂
  • MDL Number:MFCD00004910
  • Molecular Weight:150.31 g/mol
  • Boiling Pt:242…243 °C
  • Melting Pt:–21 °C
  • Density:0.990
  • Flash Pt:90 °C (194 °F)
  • Storage Temperature:Ambient
  • EINECS:214-735-1
  • UN:3334
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications