Order Entry
Italy
Orders LinkContactUsLinkComponent
Propidium Iodide

Propidium Iodide

Glass bottle, 1 mL
Catalog # TREV4830-01-03
Irritant
Marketsource
Supplier:   TREVIGEN
Propidium Iodide
Catalog # TREV4830-01-03
Supplier:  TREVIGEN
Marketsource
CAS Number:  25535-16-4
Propidium Iodide

Glass bottle, 1 mL

CAS Number:  25535-16-4
Price
Inquire For Price
Log in for availability
1 * 1 mL
Product Details & Documents
Synonyms:  3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide, PI, 3,8-Diamino-5-(3-(diethyl(methyl)ammonio)propyl)-6-phenylphenanthridin-5-ium iodide, 3,8-Diamino-5-(3-diethylaminopropyl)-6-phenylphenanthridinium iodide methiodide
  • CAS Number:25535-16-4
  • Formula:C₂₇H₃₄I₂N₄
  • MDL Number:MFCD00011921
  • Molecular Weight:668.4 g/mol
  • EINECS:247-081-0
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL

Specifications