Order Entry
Italy
Orders LinkContactUsLinkComponent
Accessories for Automatic Capillary Viscometers, ViscoSystem® AVC 370
Accessories for Automatic Capillary Viscometers, ViscoSystem® AVC 370
VZ 8571 Instrument support for CT 72/2 and CT 72/4, required for measuring systems AVS®370 in case of short hoses between ViscoPump and viscometer, e.g. with hose set VZ 8523
Catalog # SCOI1057903
Supplier:   SI Analytics
Accessories for Automatic Capillary Viscometers, ViscoSystem® AVC 370
Catalog # SCOI1057903
Supplier:  SI Analytics
Accessories for Automatic Capillary Viscometers, ViscoSystem® AVC 370
Accessories for Automatic Capillary Viscometers, ViscoSystem® AVC 370
VZ 8571 Instrument support for CT 72/2 and CT 72/4, required for measuring systems AVS®370 in case of short hoses between ViscoPump and viscometer, e.g. with hose set VZ 8523
Price
593,00 €
Log in for availability
1 * 1 pezzi
Specifications
  • Description:
    VZ 8571 Instrument support for CT 72/2 and CT 72/4, required for measuring systems AVS®370 in case of short hoses between ViscoPump and viscometer, e.g. with hose set VZ 8523
  • Seq:
    18