Sample vials, trident vials, squat or tall
Tall, 1,75 ml, 11×36 mm, Yes
Catalog # 212-7071
Supplier: MURRAY S
Jump to:
Product Details & Documents
Vials made from neutral glass with PE plug stoppers. Suitable for a wide variety of uses in the laboratory, production, more.
- Robust and economical, will withstand moderate temperature change
- Wide opening for easy filling and emptying
- Plug stopper for secure closure
Packaging: For production related reasons, the number of vials per box may vary.
Specifications
- Cap included:Yes
- Ø×H:11×36 mm
- Capacity:1,75 ml
- Description:Tall
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..



