Fluroxypyr 100 ng/µl 1 * 1 mL
Catalog # EHERXA13849000AL
Supplier: EHRENSTORFER
Frequently Bought Together
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Conductivity standards, Certipur® Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Ivermectine 100 ng/µl 1 * 1 mL
Supplier: EHRENSTORFER
Propamocarb free base 10 ng/µl 1 * 10 mL
Supplier: EHRENSTORFER