Order Entry
Italy
Orders LinkContactUsLinkComponent
DL-α-Tocopherol acetate, liquid

DL-α-Tocopherol acetate, liquid

Glass bottle for solids, 100 g
Catalog # ICNA0210055980
Supplier:   MP Biomedicals
DL-α-Tocopherol acetate, liquid
Catalog # ICNA0210055980
Supplier:  MP Biomedicals
CAS Number:  7695-91-2
DL-α-Tocopherol acetate, liquid

Glass bottle for solids, 100 g

CAS Number:  7695-91-2
Price
179,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Tocopherol acetate has properties similar but not identical to α-tocopherol.

Tocopherols (TCP) (vitamin E) are a series (α, β, γ and δ) of chiral organic molecules that vary in their degree of methylation of the phenol moiety of the chromanol ring. Tocopherols are lipid soluble anti-oxidants that protect cell membranes from oxidative damage. α-Tocopherol is the form of tocopherol preferentially absorbed by homo sapiens.

Freely soluble in absolute ethanol, acetone, chloroform (100 mg/ml - clear solution), ether and fixed oils, soluble in ethanol (96%); slightly soluble in alcohol; practically insoluble in water.

Synonyms:  (±)-α-Tocopherol acetate, all-rac-α-Tocopheryl acetate, Vitamine E acetate, DL-alpha-Tocopheryl acetate, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-yl acetate
  • CAS Number:7695-91-2
  • Formula:C₃₁H₅₂O₃
  • MDL Number:MFCD00072042
  • Molecular Weight:472.75 g/mol
  • EINECS:231-710-0

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.