Homogeniser, Dounce Type
2 ml, 60 mm
Catalog # BELC1984-10002
Marketsource
Supplier: BELLCO GLASS
Jump to:
Product Details & Documents
Dounce tissue homogeniser designed for disruption of whole cells leaving cell nuclei undamage.
- Polished tube and pestles
- Made up of glass
Delivery information: Two pestles supplied with each unit, large clearance pestle A for initial processing and small clearance pestle B completes homogenisation.
Specifications
- Capacity:2 ml
- Seq:01
- Total length:60 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



